Learn More
Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Tube Adapters for Round Rotor Buckets
For use with four-place rotors 75006445 and 75006434 when equipped with round bucket 75006441
Brand:  Thermo Scientific™ 75008136
 View more versions of this product
Product Code. 10259301
  • 165.83 € / Each
Explore more special offers
Description
Description
Adapters sold individually; order correct quantity to equip your rotor.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LH-4000W and LH-4000 rotor with round bucket
White
10/15 mL