Learn More
Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Tube Adapters for Round Rotor Buckets
For use with four-place rotors 75006445 and 75006434 when equipped with round bucket 75006441
Brand: Thermo Scientific™ 75008137
This item is not returnable.
View return policy
Description
Adapters sold individually; order correct quantity to equip your rotor.
Specifications
LH-4000W and LH-4000 rotor with round bucket | |
Olive Brown | |
15 mL |