Learn More
Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Tube Adapters for Round Rotor Buckets
For use with four-place rotors 75006445 and 75006434 when equipped with round bucket 75006441
Brand:  Thermo Scientific™ 75008140
162.31 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
 View more versions of this product
Product Code. 10395472
  • 177.68 € / Each
Explore more special offers
Description
Description
Adapters sold individually; order correct quantity to equip your rotor.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LH-4000W and LH-4000 rotor with round bucket
4 x 50 mL Bottle
Green
50 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Tube Adapters for Round Rotor Buckets > Adapter,4x50 mL Tubes