Learn More
Thermo Scientific™ Sorvall™ Lid for Primo™ Round Buckets
Provides biocontainment protection
Brand:  Thermo Scientific™ 75007598
51.85 EUR valid until 2024-11-30
Use promo code "21681" to get your promotional price.
Product Code. 10148021
  • 57.89 € / Each
Explore more special offers
Description
Description
Provides biocontainment protection
Lids fit Round Buckets (75-007-555) in 4-Place Swinging Bucket Rotor (75-007-591). Sold individually; order correct quantity for rotor.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Thermo Scientific™ Sorvall™ swing bucket