Learn More
Thermo Scientific™ Sorvall ST4F Plus Centrifuge Series
GREENER_CHOICE
Thermo Scientific Sorvall ST4F Plus Centrifuge Series
Brand:  Thermo Scientific™ 75009956
10197.00 EUR valid until 2024-12-31
Use promo code "21629" to get your promotional price.
 View more versions of this product
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 17150609
  • 11457.00 € / Each
Explore more special offers
Description
Description
The ST4F/ST4FR Plus series floor standing centrifuges come standard with an easy-to-use keypad and high contrast LCD display. This high-capacity and versatile unit offers consistent performance and is ideal for routine applications. Quickly and securely swap between 13 unique rotor options with the tool-less Auto-Lock rotor exchange. This compact floor standing unit is on casters and can easily be moved for cleaning or storage.
  • Easy to Use:
    • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance.
    • Modern industrial design supports continuous improvement in rotor loading ergonomics, user safety and current needed certifications
    • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches.
    Thermo Scientific™ Auto-Lock™ III Rotor System:
    • Secure locking system allows easy push-button installation and exchange of rotors.
    • System saves time when switching between applications.
    • Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.
    Thermo Scientific™ ClickSeal™ Bucket Sealing System:
    • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips.
    • System has glove-friendly, one-handed open/close capability.
    • System is certified by CAMR™ in Porton Down, UK.
    Thermo Scientific™ SMARTSpin™ Technology:
    • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option:
    • Technology optimizes safety and improves separations.
    Space-Saving Design:
    • Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning.
    Energy Efficient:
    • System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.
  • TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rotor needs to be purchased separately
Microprocessor
4 x 1000 mL with TX-1000 Rotor
No
No
32.8 x 22.3 x 27.2 in. (83.5 x 56.6 x 69 cm)
Up to 100 programs via push button and high contrast LCD interface
General Purpose Centrifuge
High Contrast LCD, Push Button
Energy efficient, less waste
General Use Only
EN/UL/IEC 61010-1, EN/UL/IEC 61010-2-020 and 61010-2-101, EN/UL/IEC 61326-1, EN/UL/IEC 61326-2-6, FCC part 15
15,200 rpm (Microliter 30 x 2 rotor)
25,830 x g (Microliter 30 x 2 Rotor)
SMARTSpin Imbalance detection, finger-pinch prevention, crash-proof construction
Direct, Brushless Induction Low Profile
9 Accel/10 Braking
208/240 V
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products