missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tips for Tecan™ and Axon™ Instruments PatchXpress Workstations
GREENER_CHOICE

Product Code. 11500205
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 960
This item is not returnable. View return policy

Product Code. 11500205

Brand: Thermo Scientific™ 902261XP

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with Tecan™ Genesis™, Freedom Evo™, Cavro™, PatchXpress Workstations

Clear pipet tips, Tecan style.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 50 μL
Filtered Filtered
Sterility Sterile
Color White
Autoclavable Autoclavable
Clean Claims DNase, RNase, and Pyrogen free
Format Rack
Tip Style Automation
For Use With (Equipment) Tecan Genesis, Tecan Freedom EVO, Cavro, patchXpress
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.