Learn More
Unity Lab Services™ Thermo Scientific™ Water Purification Validation Service for Thermo Scientific™ Barnstead MicroPure, all models
Stay compliant with our Industry-Exclusive, No-Charge Requalification Guarantee included with each Thermo Scientific Water Purification Validation Service for Thermo Scientific Barnstead MicroPure, all models
Brand: Unity Lab Services™ IOQPCKE50133913
1850.45 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
IQ
- Unpacking, Assembly and Installation Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order. Verifies equipment is assembled and installed and any initial diagnostics tests have been performed. Any abnormal event(s) observed during assembly and installation have been documented.
- Utility, Facility and Environment Verifies that the installation site satisfactorily meets manufacturer-specified environmental requirements.
OQ
- System Component Information Detailed configuration information for each system component have been recorded.
- OQ Limits: Record a list of manufacturer recommended limits for ensuring that the system is operating as expected.
- Equipment Operational Tests and Results: Test important equipment functions to verify that the equipment operates as intended by the manufacturer and required by the user. This is a group of selected important equipment parameters for testing according to the intended use of the equipment.
Key Tests: Flow Adjustment, Water Pressure, Resistivity, Power Failure
Specifications
Thermo Scientific Water Purification including Thermo Scientific Barnstead MicroPure, all models | |
Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, Flow Adjustment, Water Pressure, Resistivity, Power Failure testing. | |
Contact your local sales representative for a list of standard and extended coverage areas. | |
Travel to major metropolitan areas. Does not include Installation. Instrument must be installed, powered-up and set temperature point for 24 hours before on-site service |