Learn More
Thermo Scientific™ Thermo Series Liquid Nitrogen Transfer Vessels
Store and dispense small amounts of liquid nitrogen with our economical and durable liquid nitrogen transfer vessels.
Brand:  Thermo Scientific™ TY509X1
1300.05 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 10480394
  • 1517.55 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Store and dispense small amounts of liquid nitrogen with our economical and durable Thermo Scientific™ Thermo Series Liquid Nitrogen Transfer Vessels. The Thermo Series includes six models with capacities from 5 to 32 L. A full range of handling and dispensing accessories as well as transport carts are also available.
Features:
  • Designed to bring liquid nitrogen to other cryovessels
  • Lightweight aluminum dewars assure low-static evaporation rates
  • Optional withdrawal device, 12mL dipper and wheeled accessory cart
  • Compact Thermo 5 and 10 dewars feature a convenient pail-style handle for pouring and for use in applications where only small quantities of liquid nitrogen are needed
  • Thermo 10, 20, and 30 can be fitted with self-pressurized withdrawal accessory devices to easily dispense LN2 without pouring
Note:
Stated static evaporation rates are based on new container performance with no lid openings and no product loads. Actual working performance may vary with individual applications.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LN2 Transfer Vessel
18.2 in.
8.8 in.
22.4 cm
12 lb.
8 lb.
5 L
46.2 cm
2.2 in.
5.6 cm
5.4 kg
3.6 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Thermo Series Liquid Nitrogen Transfer Vessels > Capacity: 5L