Learn More
Eppendorf™ ThermoMixer
Brand:  Eppendorf™ 5385000032
2650.15 EUR valid until 2024-12-06
Use promo code "21633" to get your promotional price.
Product Code. 15109762
  • 3350.00 € / Each
Explore more special offers
Specifications
±0.5°C at 20° to 45°C
Thermoblock For Microplates and Deep Well Plates, Including Lid
220 to 240 V, 50/60 Hz, 200 W
170 mm
304 mm
50 to 60 Hz
200 W
4 to 100°C
220 to 240 V
ThermoMixer
206 mm
Transformation of Bacteria Strains and Plasmids
4°C to 100°C
2000 rpm