missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Deep Well Plate Thermoshaker TS-DW

A thermoshaker designed for shaking and heating of deep well plates. A multisystem principle, used in the design of the thermoshaker, allows it to operate as 3 independent devices: Incubator, Plate shaker and Thermoshaker. Accessory blocks available separately.

Brand:  Grant Instruments™ TS-DW

Product Code. 15420474

  • 1740.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Temperature range 25°C to 100°C
  • Temperature control, ambient 5°C to 100°C
  • Platform heat-up speed from 25°C to 37°C: 6 minutes (For B-2E block)
  • Shaking speed 250 to 1400 rpm
  • Profiled platform to fit perfectly with your block, allowing maximum heat transfer
  • Small footprint
  • Mixing and incubation are combined to reduce reaction process times
  • Choice of 4 interchangeable profiled blocks are available for specific deep well plates
  • 2-line LCD display indicates both set and actual values
  • Heated lid supplied as standard
  • Programmable with digital timer (with sound alarm)
Note: Blocks are sold separately.
Specifications

Specifications

250 to 1400 rpm
25°C to 100°C
2 mm
5.0 kg
ELISA, PCR and molecular biology prep, thawing/mixing, master mix preparation, life science, cell biology, cell lysis, DNA/RNA isolation and purification, and pellet re-suspension
Deep Well Plate Thermoshaker, Heated Lid. (Blocks are sold separately.)
Input AC 100/240 V, 50/60 Hz, Output DC 12 V
0.1°C (at 37°C)
260 x 240 x 130 mm
0.5°C (at 37°C)
1 deep well plate, Eppendorf, Sarstedt, Axygen, Starlab, Custom (must be ordered separately)
1 min. to 96 hr.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.