missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thiamine hydrochloride, 98.5-101.5%

Thiamine hydrochloride, 98.5-101.5%, C12H17ClN4OS.HCl, CAS Number-67-03-8, aneurine hydrochloride, berin, thiamin chloride, thiamine chloride hydrochloride, thiamine dichloride, thiamine hcl, thiamine hydrochloride, thiaminium chloride, trophite, vitamin b1 hydrochloride, 1kg, 98.5 to 101.5%

28.84 € - 694.46 €

Chemical Identifiers

CAS 67-03-8
Molecular Formula C12H17ClN4OS·HCl
Molecular Weight (g/mol) 337.26
MDL Number MFCD00012780
InChI Key DPJRMOMPQZCRJU-UHFFFAOYSA-M
Synonym thiamine hydrochloride, vitamin b1 hydrochloride, thiamine hcl, aneurine hydrochloride, berin, thiamine chloride hydrochloride, trophite, thiamin chloride, thiamine dichloride, thiaminium chloride
PubChem CID 6202
ChEBI CHEBI:49105
IUPAC Name 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
SMILES CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10477540
SDS View Documents
Thermo Scientific Chemicals
148990100
10 g Glass bottle
28.84 €
10g
Please to purchase this item. Need a web account? Register with us today!
10697492
SDS View Documents
Thermo Scientific Chemicals
148991000
100 g Glass bottle
131.66 €
100g
Please to purchase this item. Need a web account? Register with us today!
10023953
SDS View Documents
Thermo Scientific Chemicals
148990010
1 kg Glass Bottle
694.46 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

67-03-8
337.26
DPJRMOMPQZCRJU-UHFFFAOYSA-M
6202
2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
C12H17ClN4OS·HCl
MFCD00012780
thiamine hydrochloride, vitamin b1 hydrochloride, thiamine hcl, aneurine hydrochloride, berin, thiamine chloride hydrochloride, trophite, thiamin chloride, thiamine dichloride, thiaminium chloride
CHEBI:49105
CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]

Specifications

67-03-8
White
98.5 to 101.5%
MFCD00012780
15, 9447
Solubility in water: 1 g/mL. Other solubilities: 1 g/18 mL glycerol - 1 g/100 mL 95% alcohol, 1 g/315 mL abs. alcohol, soluble in propylene glycol, practically insoluble in hexane, chloroform, insoluble in benzene, diethyl ether
CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]
337.26
CHEBI:49105
98.5-101.5%
Thiamine hydrochloride, 98.5-101.5%
250.0°C
Authentic
C12H17ClN4OS·HCl
27, IV, 1766
thiamine hydrochloride, vitamin b1 hydrochloride, thiamine hcl, aneurine hydrochloride, berin, thiamine chloride hydrochloride, trophite, thiamin chloride, thiamine dichloride, thiaminium chloride
DPJRMOMPQZCRJU-UHFFFAOYSA-M
2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
6202
337.26
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 200-641-8

missing translation for 'rtecsNumber' : XI7350000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.