Learn More
Thermo Scientific Chemicals Thiazolyl Blue tetrazolium bromide, 98%
Description
Used in measurement of cell proliferation. Produces a yellowish solution that is converted to dark blue, water-insoluble MTT formazan by mitochondrial dehydrogenases of living cells.Thiazolyl Blue tetrazolium bromide is a dye used in measurement of cell proliferation. It is used as a direct indicator of cytotoxicity (such as for screening cancer drugs) and apoptosis. As an electron acceptor, it is utilized for studying nicotinamide adenine dinucleotide phosphate (NADP)-dependent dehydrogenases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used in measurement of cell proliferation. Produces a yellowish solution that is converted to dark blue, water-insoluble MTT formazan by mitochondrial dehydrogenases of living cells.Thiazolyl Blue tetrazolium bromide is a dye used in measurement of cell proliferation. It is used as a direct indicator of cytotoxicity (such as for screening cancer drugs) and apoptosis. As an electron acceptor, it is utilized for studying nicotinamide adenine dinucleotide phosphate (NADP)-dependent dehydrogenases.
Solubility
Soluble in water, ethanol, 2-methoxyethanol and phosphate buffered saline.
Notes
Light, air and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | ∽195°C (decomposition) |
Odor | Odorless |
Quantity | 1 g |
Beilstein | 3825277 |
Sensitivity | Air, light, and moisture sensitive |
Solubility Information | Soluble in water,ethanol,2-methoxyethanol and phosphate buffered saline. |
Formula Weight | 414.33 |
Percent Purity | 98% |
Chemical Name or Material | Thiazolyl Blue tetrazolium bromide |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z31K003 | Certificate of Analysis | 06/01/2025 | L11939.03,L11939.06 |
S13K046 | Certificate of Analysis | 03/07/2024 | L11939.03,L11939.06 |
N28K024 | Certificate of Analysis | 05/03/2024 | L11939.03,L11939.06 |
S08J050 | Certificate of Analysis | 16/06/2023 | L11939.03,L11939.06,L11939 |
10246277 | Certificate of Analysis | 09/05/2023 | L11939.03,L11939.06 |
Safety and Handling

- Thiazolyl Blue tetrazolium bromide
- Warning
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.