missing translation for 'onlineSavingsMsg'
Learn More
Thieno[2,3-b]pyridine-2-carboxylic acid, 95%, Thermo Scientific™ Product Code.: 10351043

Thieno[2,3-b]pyridine-2-carboxylic acid, 95%, Thermo Scientific™

Product Code. 10351043
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10351043

Brand: Thermo Scientific Maybridge CC05601CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 59944-76-2
Molecular Formula C8H5NO2S
Molecular Weight (g/mol) 179.193
MDL Number MFCD07371381
InChI Key XGCSHAYMNOFFNA-UHFFFAOYSA-N
Synonym thieno 2,3-b pyridine-2-carboxylic acid, thieno 2,3-b pyridine-2-carboxylicacid, 2-carboxythieno 2,3-b pyridine, thiopheno 2,3-b pyridine-2-carboxylic acid, hieno 2,3-b pyridine-2-carboxylic acid
PubChem CID 7162095
IUPAC Name thieno[2,3-b]pyridine-2-carboxylic acid
SMILES C1=CC2=C(N=C1)SC(=C2)C(=O)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 179.2
Percent Purity 95%
Chemical Name or Material Thieno[2, 3-b]pyridine-2-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thieno[2,3-b]pyridine-2-carboxylic acid, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.