missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thieno[3,2-b]pyridin-7-ol, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Thieno[3,2-b]pyridin-7-ol is used for pharmaceutical intermediates, as drugs, food, cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thieno[3,2-b]pyridin-7-ol is used for pharmaceutical intermediates, as drugs, food, cosmetics.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Thieno[3,2-b]pyridin-7-ol is used for pharmaceutical intermediates, as drugs, food, cosmetics.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Quantity | 1 g |
Solubility Information | Soluble in water. |
Formula Weight | 151.19 |
Percent Purity | 96% |
Chemical Name or Material | Thieno[3,2-b]pyridin-7-ol |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thieno[3,2-b]pyridin-7-ol, 96%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction