missing translation for 'onlineSavingsMsg'
Learn More

Thieno[3,2-b]pyridine-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10738455
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 347187-30-8
Molecular Formula C8H5NO2S
Molecular Weight (g/mol) 179.193
MDL Number MFCD12198115
InChI Key WVWCTPQHJWMLKI-UHFFFAOYSA-N
Synonym thieno 3,2-b pyridine-2-carboxylic acid, thiopheno 3,2-b pyridine-2-carboxylic acid
PubChem CID 22047169
IUPAC Name thieno[3,2-b]pyridine-2-carboxylic acid
SMILES C1=CC2=C(C=C(S2)C(=O)O)N=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 179.2
Percent Purity 97%
Chemical Name or Material Thieno[3, 2-b]pyridine-2-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.