missing translation for 'onlineSavingsMsg'
Learn More

Wilmad-LabGlass™ Thin Walled High Throughput NMR Tubes

Product Code. 15329038 Shop All SP Scienceware Products
Click to view available options
Length (Metric):
103.5 mm
178 mm
203 mm
Field Strength:
600 MHz
High-throughput
Quantity:
100/Pk.
50/Pk.
Unit Size:
Pack of 100
Pack of 50
This item is not returnable. View return policy

Product Code. 15329038

Brand: Wilmad-LabGlass™ 669995800

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Possess an average camber of 60 microns to guarantee the clarity of the spectrum for small molecule (MW<250) samples up to 600 MHz. Wilmad Thin Walled High Throughput NMR Tubes are designed for routine use in most low to mid field NMR spectrometers.

  • Camber and concentricity values listed are the Total Indicator Reading (TIR)
  • Designed for routine use in most low to mid field NMR spectrometers
  • One of the best OD tolerances in the industry
  • Guaranteed to fit tightly in spinner turbine
  • Made from ASTM E438 Type 1 Class B glass
  • 100% inspected for surface defects and physical dimension to ensure the success of your experiments
  • WG-1000-4-SJ and WG-1000-7-SJ are supplied with authentic Bruker SampleJet caps, all others come with a disposable NMR cap
Note: For NMR experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, please refer to Wilmad Precision NMR Tubes.

Specifications

Camber (Metric) 60 μm
Length (Metric) 203 mm
Material ASTM E438 Type 1 Class B Glass
Product Type NMR Tube
Closure Type Disposable NMR Cap
Diameter (Metric) Outer 4.94665±0.01905 mm
Field Strength High-throughput
Length (English) 8 in.
Thickness (Metric) Wall 0.43 mm
Quantity 100/Pk.
Model WG-1000-8
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.