missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Thinline Biopsy Cassettes in Tube Packs

For biopsy tissue samples, cassettes have a front lid hinge, attached lids in matching colors, and are compatible with all Epredia PrintMate™ Cassette Printers

224.00 €

Specifications

Closure Included Yes
Quantity 750/Cs.
Pore Style Small Squares
Closure Type Attached Lid, Front Hinge
PrintMate Compatible Yes
View More Specs

Products 10
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
16304116
View Documents
Epredia™
B851737BE
Beige
224.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16314116
View Documents
Epredia™
B851737BL
Blue
224.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16324116
View Documents
Epredia™
B851737GN
Green
224.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16334116
View Documents
Epredia™
B851737GY
Gray
224.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16354116
View Documents
Epredia™
B851737LG
Light Green
224.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16364116
View Documents
Epredia™
B851737LI
Lilac
224.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16374116
View Documents
Epredia™
B851737OR
Orange
224.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16384116
View Documents
Epredia™
B851737PK
Pink
224.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16394116
View Documents
Epredia™
B851737TN
Tan
224.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16304126
View Documents
Epredia™
B851737WH
White
224.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing, Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving
Specifications

Specifications

Yes
Small Squares
Yes
Thinline Biopsy Cassette
750/Cs.
Attached Lid, Front Hinge
For Use With PrintMate™ Printers
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.