Learn More
Thioacetamide, 98%
Description
Thioacetamide is used in qualitative inorganic analysis as an in-situ source for sulfide ions. It is used in analytical chemistry as a source of hydrogen sulfide. It is an additive in enantioselective reduction of beta-keto esters with immobilized bakers yeast. It is also used as a vulcanizing agent of polymer and a crosslinking agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Thioacetamide is used in qualitative inorganic analysis as an in-situ source for sulfide ions. It is used in analytical chemistry as a source of hydrogen sulfide. It is an additive in enantioselective reduction of beta-keto esters with immobilized bakers yeast. It is also used as a vulcanizing agent of polymer and a crosslinking agent.
Solubility
Soluble in ethanol, benzene and petroleum ether. Slightly soluble in water and ether.
Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
Specifications
Specifications
| Melting Point | 110°C to 114°C |
| Density | 1.37 |
| Assay Percent Range | 98% |
| Quantity | 500 g |
| Beilstein | 506006 |
| Merck Index | 14,9319 |
| Solubility Information | Soluble in ethanol,benzene and petroleum ether; Slightly soluble in water and ether. |
| Formula Weight | 75.13 |
| Percent Purity | 98% |
| Physical Form | Crystalline |
| Show More |
Safety and Handling
- Thioacetamide
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H350-May cause cancer.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Thioacetamide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.