Learn More
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 112.0°C to 114.0°C |
Color | White to Beige |
Density | 1.3700g/mL |
Ignition Residue | 0.05% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (Argentometry) |
Packaging | Glass bottle |
Linear Formula | CH3CSNH2 |
Quantity | 100 g |
Beilstein | 02, 232 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0452616 | Certificate of Analysis | 01/05/2024 | 424535000,424531000,424530250 |
A0444853 | Certificate of Analysis | 25/08/2023 | 424535000,424531000,424530250 |
A0402854 | Certificate of Analysis | 13/12/2022 | 424535000,424531000,424530250 |
A0431348 | Certificate of Analysis | 21/06/2022 | 424535000,424531000,424530250 |
A0426203 | Certificate of Analysis | 20/01/2022 | 424535000,424531000,424530250 |
Safety and Handling


- Thioacetamide
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H350-May cause cancer.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.