missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiocarbanilide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Plastic Drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 151.0°C to 154.0°C |
Color | White to Yellow |
Loss on Drying | 0.3% max. (60°C) |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Plastic bottle |
Linear Formula | C6H5NHCSNHC6H5 |
Quantity | 100 g |
Beilstein | 12,394 |
Merck Index | 15,3373 |
Show More |
Safety and Handling
Product Identifier
- Thiocarbanilide
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H317-May cause an allergic skin reaction.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'