missing translation for 'onlineSavingsMsg'
Learn More

Thiomorpholine, 98%, Thermo Scientific Chemicals

Product Code. 11468516
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Thiomorpholine is used as an intermediate for blonanserin. It is used in the preparation of pyrrole derivatives, which acts as a antimycobacterial agent. It is further involved in the hypocholesterolemic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiomorpholine is used as an intermediate for blonanserin. It is used in the preparation of pyrrole derivatives, which acts as a antimycobacterial agent. It is further involved in the hypocholesterolemic activity.

Solubility
Miscible with waterMiscible with water and organic solvents.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong bases and air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-90-0
Molecular Formula C4H9NS
Molecular Weight (g/mol) 103.183
MDL Number MFCD00005974
InChI Key BRNULMACUQOKMR-UHFFFAOYSA-N
Synonym thiamorpholine, thiazolidinane, parathiazan, 4-thiomorpholine, tetrahydro-1,4-thiazine, 1,4-thiazane, 1,4-thiazan, 1-thia-4-azacyclohexane, unii-3a8r61g6qv, 1,4-thiazaperhydroine
PubChem CID 67164
ChEBI CHEBI:36392
IUPAC Name thiomorpholine
SMILES C1CSCCN1

Specifications

Density 1.086
Boiling Point 175°C to 177°C
Flash Point 67°C (153°F)
Refractive Index 1.5384
Quantity 25 g
UN Number UN3267
Beilstein 102550
Sensitivity Air Sensitive
Merck Index 14,9300
Solubility Information Miscible with waterMiscible with water and organic solvents.
Formula Weight 103.19
Percent Purity 98%
Chemical Name or Material Thiomorpholine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'