missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Thionin acetate, pure, high purity biological stain
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator

Specifications
Specifications
Dye Content | 85% min. |
Beilstein | 27, 392 |
Merck Index | 14, 9346 |
Solubility Information | Solubility in water: ca 2.5g/L (25°C). Other solubilities: practically insoluble in ethanol |
Infrared Spectrum | Authentic |
Lambda Max | 597 to 601nm (in water) |
Formula Weight | 287.33 |
Grade | Pure |
Packaging | Glass bottle |
UV Spectrometry | Complies |
Show More |
Safety and Handling

Product Identifier
- Thionin acetate, high purity biological stain
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Thionin acetate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction