missing translation for 'onlineSavingsMsg'
Learn More
Thiophene-2-thiol, contains dimer, 97% as monomer, Thermo Scientific Chemicals Product Code.: 11415257

Thiophene-2-thiol, contains dimer, 97% as monomer, Thermo Scientific Chemicals

Product Code. 11415257
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11415257

Brand: Thermo Scientific Alfa Aesar B22642.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7774-74-5
Molecular Formula C4H4S2
Molecular Weight (g/mol) 116.196
MDL Number MFCD00051666
InChI Key SWEDAZLCYJDAGW-UHFFFAOYSA-N
Synonym 2-mercaptothiophene, 2-thiophenethiol, thiophenethiol, 2-thienyl mercaptan, 2-thienylthiol, 2-thiophene thiol, unii-f3lb1437xq, 2-sulphanylthiophene, alpha-thienyl mercaptan, thiophene thiol
PubChem CID 522674
IUPAC Name thiophene-2-thiol
SMILES C1=CSC(=C1)S

Specifications

Density 1.247
Boiling Point 62°C to 65°C (12 mmHg)
Flash Point 66°C (151°F)
Odor Unpleasant
Refractive Index 1.617
Quantity 5 g
Beilstein 104650
Sensitivity Air and light sensitive
Formula Weight 116.2
Percent Purity 97%
Assay as monomer, contains dimer
Chemical Name or Material Thiophene-2-thiol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thiophene-2-thiol, contains dimer, 97% as monomer, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.