Learn More
Thiophene-3-carbonitrile, 98%, Thermo Scientific Chemicals
57.50 € - 188.00 €
Chemical Identifiers
CAS | 1641-09-4 |
---|---|
Molecular Formula | C5H3NS |
Molecular Weight (g/mol) | 109.146 |
MDL Number | MFCD00151852 |
InChI Key | GSXCEVHRIVLFJV-UHFFFAOYSA-N |
Synonym | 3-cyanothiophene, 3-thiophenecarbonitrile, thiophene-3-nitrile, pubchem5508, pubchem12351, acmc-209dqk, ksc176g4h |
PubChem CID | 74231 |
IUPAC Name | thiophene-3-carbonitrile |
SMILES | C1=CSC=C1C#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11337906
|
Thermo Scientific Alfa Aesar
L13115.06 |
5 g |
57.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11347906
|
Thermo Scientific Alfa Aesar
L13115.14 |
25 g |
188.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1641-09-4 | |
109.146 | |
GSXCEVHRIVLFJV-UHFFFAOYSA-N | |
74231 | |
C1=CSC=C1C#N |
C5H3NS | |
MFCD00151852 | |
3-cyanothiophene, 3-thiophenecarbonitrile, thiophene-3-nitrile, pubchem5508, pubchem12351, acmc-209dqk, ksc176g4h | |
thiophene-3-carbonitrile |
Specifications
1641-09-4 | |
205°C | |
Recognizable | |
1.5615 | |
UN3276 | |
3-cyanothiophene, 3-thiophenecarbonitrile, thiophene-3-nitrile, pubchem5508, pubchem12351, acmc-209dqk, ksc176g4h | |
C1=CSC=C1C#N | |
109.146 | |
109.15 | |
Thiophene-3-carbonitrile |
1.2 | |
86°C (186°F) | |
C5H3NS | |
MFCD00151852 | |
107104 | |
GSXCEVHRIVLFJV-UHFFFAOYSA-N | |
thiophene-3-carbonitrile | |
74231 | |
98% |
Safety and Handling
GHS H Statement
H301-H227-H312-H332-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c
H227-H302+H312+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 216-687-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only