missing translation for 'onlineSavingsMsg'
Learn More

Thiosalicylic acid, 98%, Thermo Scientific Chemicals

Product Code. 10173470
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-93-3
Molecular Formula C7H6O2S
Molecular Weight (g/mol) 154.183
MDL Number MFCD00004836
InChI Key NBOMNTLFRHMDEZ-UHFFFAOYSA-N
Synonym thiosalicylic acid, 2-mercaptobenzoic acid, o-mercaptobenzoic acid, 2-thiosalicylic acid, o-thiosalicylic acid, 2-carboxythiophenol, o-benzoic acid thiol, o-carboxythiophenol, o-sulfhydrylbenzoic acid, benzoic acid, 2-mercapto
PubChem CID 5443
ChEBI CHEBI:59124
IUPAC Name 2-sulfanylbenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)S

Specifications

Melting Point 164.0°C to 169.0°C
Color White to Yellow
Assay Percent Range 98%
Linear Formula HSC6H4CO2H
Beilstein 10, 125
Merck Index 15, 9513
Solubility Information Solubility in water: soluble in hot water. Other solubilities: freely soluble in acetic acid, soluble in alcohol and ether, soluble 200 mg/l in ethanol
Formula Weight 154.18
Percent Purity 98%
Quantity 100 g
Flash Point 150°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Fine Crystalline Powder
Chemical Name or Material Thiosalicylic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Thiosalicylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Thiosalicylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'