missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiourea, 99+%, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 171.0°C to 175.0°C |
Color | White |
Ignition Residue | 0.1% max. |
Loss on Drying | 0.5% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (On dried basis) (Argentometry) |
Packaging | Plastic bottle |
Linear Formula | H2NCSNH2 |
Quantity | 250 g |
Beilstein | 03, 180 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0457631 | Certificate of Analysis | 08/11/2023 | 424542500,424540010 |
A0453261 | Certificate of Analysis | 25/08/2023 | 424542500,424540010 |
A0393546 | Certificate of Analysis | 14/12/2022 | 424542500,424540010 |
A0413821 | Certificate of Analysis | 13/12/2022 | 424542500,424540010 |
A0400012 | Certificate of Analysis | 12/12/2022 | 424542500,424540010 |
Safety and Handling



Product Identifier
- Thiourea
- Warning
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- H351-Suspected of causing cancer.
- H411-Toxic to aquatic life with long lasting effects.
- H361d-Suspected of damaging the unborn child.
- H302-Harmful if swallowed.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P310-Immediately call a POISON CENTER/doctor/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P273-Avoid release to the environment.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thiourea, 99+%, ACS reagent, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction