missing translation for 'onlineSavingsMsg'
Learn More

Thirty three elements solution for ICP 100mg/L HNO3 matrix, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Brand:  Fisher Chemical MS 101 500


 View more versions of this product

Product Code. 12936213

  • 575.00 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thirty three elements solution - Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Cs, Co, Cr,Cu, Fe, In, K, Li, Mg, Mn, Mo, Na, Nb, Ni, Pb,Rb Sb Se Sr, Ti, Tl, V, U, Zn for ICP 100mg/L HNO3 matrix
Specifications

Specifications

Thirty Three Elements Solution
Bottle
ICP Multi-element Standard Solution
Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Cs, Co, Cr,Cu, Fe, In, K, Li, Mg, Mn, Mo, Na, Nb, Ni, Pb,Rb, Sb, Se, Sr, Ti, Tl, V, U, Zn
Liquid
100mg/L
500 mL
1760
5% HNO3
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Multi-element Standard Solution for ICP. 33 Components in HNO3 5% (MS 101)

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Carcinogenicity Category 1B
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H350-May cause cancer.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Certificates
Special Offers

Special Offers