Learn More
SCAT™ PTFE Thread Adapters
SCAT™ Safety Caps and SCAT™ Safety Waste Caps are available in many thread sizes. If the right thread for your container is still not available, simply use the matching adapter.
Brand:  SCAT™ 117094
67.50 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10840411
  • 85.40 € / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
SCAT™ thread adapters are ideally suited for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
  • Made of polytetrafluoroethylene (PTFE) material
  • Available in a wide range of thread sizes
  • Suitable for all SCAT™ caps, funnels or canisters
Specifications
Specifications
GL45 (f) to S55 (m)
PTFE
Thread Adapter