missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Three-Neck Round-Bottom Flasks, Side Necks Angled

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

62.00 € - 352.00 €

Specifications

Standard Taper Joint NS 29/32 Center Neck
View More Specs

Products 5
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10119360
View Documents
Lenz Laborglasintrumente
03314170
1 L
62.00 €
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
 
10290921
View Documents
Lenz Laborglasintrumente
03314173
2 L
94.00 €
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
 
10283901
View Documents
Lenz Laborglasintrumente
03314179
4 L
283.00 €
Each
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
 
10315171
View Documents
Lenz Laborglasintrumente
03314185
6 L
352.00 €
Each
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
 
10726951
View Documents
Lenz Laborglasintrumente
03314176
3 L
252.00 €
Each
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
 
Description

Description

  • Distillation flasks acc. to DIN 12394 and in addition to DIN standard
  • With two angled side necks (20°)
  • With conical ground joints, with inscription label
  • Material: DURAN™
Specifications

Specifications

NS 29/32 Center Neck
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lenz Laborglasintrumente Three-Neck Round-Bottom Flasks, Side Necks Angled

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.