missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) bromide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

144.72 € - 631.14 €

Chemical Identifiers

CAS 14456-51-0
Molecular Formula Br3Tm
Molecular Weight (g/mol) 408.646
MDL Number MFCD00050138
InChI Key HQSWGSFQSCMHFQ-UHFFFAOYSA-K
Synonym thulium bromide, thulium tribromide, thulium iii bromide, thulium 3+ tribromide, acmc-1bo43, thulium iii bromide, anhydrous, ampuled under argon trace metals basis 1g
PubChem CID 84454
IUPAC Name thulium(3+);tribromide
SMILES [Br-].[Br-].[Br-].[Tm+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11353568
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
039186.03
1 g
144.72 €
1g
Please to purchase this item. Need a web account? Register with us today!
11343568
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
039186.06
5 g
631.14 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

14456-51-0
408.646
HQSWGSFQSCMHFQ-UHFFFAOYSA-K
84454
[Br-].[Br-].[Br-].[Tm+3]
Br3Tm
MFCD00050138
thulium bromide, thulium tribromide, thulium iii bromide, thulium 3+ tribromide, acmc-1bo43, thulium iii bromide, anhydrous, ampuled under argon trace metals basis 1g
thulium(3+);tribromide

Specifications

1440°C
14456-51-0
Br3Tm
thulium bromide, thulium tribromide, thulium iii bromide, thulium 3+ tribromide, acmc-1bo43, thulium iii bromide, anhydrous, ampuled under argon trace metals basis 1g
[Br-].[Br-].[Br-].[Tm+3]
408.646
408.66
Ampule
(REO)
952°C
Powder
MFCD00050138
HQSWGSFQSCMHFQ-UHFFFAOYSA-K
thulium(3+);tribromide
84454
99.99%
Hygroscopic
Thulium(III) bromide, Ultra dry
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 238-444-4

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only