missing translation for 'onlineSavingsMsg'
Learn More
Thulium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11336238

Thulium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11336238
10 g
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy

Product Code. 11336238

Brand: Thermo Scientific Alfa Aesar 011200.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thulium(III) chloride hydrate is used in proteomics research. It is also used in the production of glass, phosphors and lasers. Further, it serves as an important dopant for fiber amplifiers. Further, it is also used to prepare thulium(III) oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19423-86-0
Molecular Formula Cl3Tm
Molecular Weight (g/mol) 275.28
MDL Number MFCD00011283
InChI Key ILOTUXNTERMOJL-UHFFFAOYSA-K
IUPAC Name thulium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Tm+3]

Specifications

Color Green
Physical Form Crystalline Aggregates
Quantity 10 g
Assay Percent Range 99.9% (REO)
Solubility Information Soluble in water and alcohol.
Formula Weight 275.29 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Thulium(III) chloride hydrate
compliance-icons
Product Identifier
  • Thulium(III) chloride hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Thulium(III) chloride hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thulium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.