Learn More
Thulium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
75.58 € - 803.00 €
Chemical Identifiers
CAS | 19423-86-0 |
---|---|
Molecular Formula | Cl3Tm |
Molecular Weight (g/mol) | 275.28 |
MDL Number | MFCD00011283 |
InChI Key | ILOTUXNTERMOJL-UHFFFAOYSA-K |
IUPAC Name | thulium(3+) trichloride |
SMILES | [Cl-].[Cl-].[Cl-].[Tm+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11346238
|
Thermo Scientific Alfa Aesar
011201.02 |
0.5 g |
75.58 €
0.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356238
|
Thermo Scientific Alfa Aesar
011201.04 |
2 g |
215.07 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11366238
|
Thermo Scientific Alfa Aesar
011201.09 |
10 g |
803.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thulium(III) chloride hydrate is used extremely as a rare metal to study the metal science of thulium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
19423-86-0 | |
275.28 | |
ILOTUXNTERMOJL-UHFFFAOYSA-K | |
[Cl-].[Cl-].[Cl-].[Tm+3] |
Cl3Tm | |
MFCD00011283 | |
thulium(3+) trichloride |
Specifications
19423-86-0 | |
Wet Crystalline Aggregates | |
Cl3Tm | |
Soluble in water. | |
[Cl-].[Cl-].[Cl-].[Tm+3] | |
275.28 | |
Odorless | |
Thulium(III) chloride hydrate |
Green | |
99.99% (REO) | |
MFCD00011283 | |
ILOTUXNTERMOJL-UHFFFAOYSA-K | |
thulium(3+) trichloride | |
275.29 (Anhydrous) | |
Hygroscopic |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 236-904-9
missing translation for 'rtecsNumber' : XP0525000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.