missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) fluoride, ultra dry, 99.99% (REO), Thermo Scientific™

Product Code. 11358937
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13760-79-7
Molecular Formula F3Tm
Molecular Weight (g/mol) 225.93
MDL Number MFCD00049611
InChI Key AATUHDXSJTXIHB-UHFFFAOYSA-K
IUPAC Name thulium(3+) trifluoride
SMILES [F-].[F-].[F-].[Tm+3]

Specifications

Boiling Point 2200°C
Melting Point 1158°C
Physical Form Powder
Quantity 1 g
UN Number UN3288
Solubility Information It is insoluble in water.
Formula Weight 225.93
Percent Purity 99.99%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 7.9 g/mL
Assay (REO)
Chemical Name or Material Thulium(III) fluoride, Ultra dry
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.