missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) nitrate hydrate, REacton™, 99.9% (REO)

Product Code. p-7049517
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11382528

Brand: Thermo Scientific Alfa Aesar 014579.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thulium(III) nitrate hydrate is used as a laboratory reagent. It finds application in optical glasses, structural ceramics, catalysts, electrical components and photo-optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00149875

Specifications

Physical Form Crystalline Aggregates
Quantity 2 g
UN Number UN1477
Solubility Information Soluble in water.
Formula Weight 354.95 (Anhydrous)
Percent Purity 99.9%
Grade Reacton
Assay (REO)
Chemical Name or Material Thulium(III) nitrate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.