missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) oxalate hydrate, REacton™, 99.9% (REO), Thermo Scientific™

Product Code. 11372528
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58176-73-1
Molecular Formula C6O12Tm2
Molecular Weight (g/mol) 601.92
MDL Number MFCD00050804
InChI Key BSCZSHAJTJRUPP-UHFFFAOYSA-H
Synonym thulium oxalate hydrate, thulium iii oxalate hydrate, acmc-20ajie, ethanedioic acid,thulium 3+ salt,hydrate 3:2:?
PubChem CID 45052048
SMILES [Tm+3].[Tm+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O

Specifications

Physical Form Crystalline
Quantity 10 g
Linear Formula Tm2(C2O4)3·xH2O
UN Number UN3288
Merck Index 14,9394
Solubility Information Soluble in aqueous alkali oxalates with double oxalate formation
IUPAC Name dithulium(3+) trioxalate
Formula Weight 601.93 (Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Assay (REO)
Chemical Name or Material Thulium(III) oxalate hydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'