missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

197.99 € - 632.15 €

Chemical Identifiers

CAS 12036-44-1
Molecular Formula O3Tm2
Molecular Weight (g/mol) 385.87
MDL Number MFCD00011285
InChI Key ZIKATJAYWZUJPY-UHFFFAOYSA-N
IUPAC Name dithulium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Tm+3].[Tm+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11316238
SDS View Documents
Thermo Scientific Alfa Aesar
011199.04
2 g
197.99 €
2g
Please to purchase this item. Need a web account? Register with us today!
11306238
SDS View Documents
Thermo Scientific Alfa Aesar
011199.09
10 g
632.15 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thulium oxide is an excellent thulium source which finds use in glass, optical and ceramic applications. Further, it is used in the manufacturing of portable X-ray transmission device. In addition to this, it finds use as a nuclear reactor control material. Nanostructured thulium oxide acts as an efficient biosensor in the field of medicinal chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 234-851-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thulium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.