missing translation for 'onlineSavingsMsg'
Learn More
Thulium(III) oxide, REacton™, 99.997% (REO), Thermo Scientific Chemicals Product Code.: 11331448

Thulium(III) oxide, REacton™, 99.997% (REO), Thermo Scientific Chemicals

Product Code. 11331448
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11331448

Brand: Thermo Scientific Alfa Aesar 010918.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thulium(III) oxide when irradiated in a nuclear reactor, thulium produces an isotope that emits x-rays. A button of this isotope is used to make a lightweight, portable x-ray machine for medical use. It is also used in lasers with surgical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12036-44-1
Molecular Formula O3Tm2
Molecular Weight (g/mol) 385.87
MDL Number MFCD00011285
InChI Key ZIKATJAYWZUJPY-UHFFFAOYSA-N
IUPAC Name dithulium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Tm+3].[Tm+3]

Specifications

Melting Point 2425°C
Color Green
Physical Form Powder
Quantity 5 g
Linear Formula Tm2O3
Merck Index 14,9394
Solubility Information Insoluble in water. Slightly soluble in acids.
Formula Weight 385.87
Percent Purity 99.997%
Grade Reacton™
Assay (REO)
Chemical Name or Material Thulium(III) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thulium(III) oxide, REacton™, 99.997% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.