missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Thulium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific™

78.00 €

Chemical Identifiers

CAS 14014-10-9
Molecular Formula Cl3O12Tm
Molecular Weight (g/mol) 467.272
MDL Number MFCD00049609
InChI Key JNNWMYZIHRZEKX-UHFFFAOYSA-K
Synonym thulium 3+ perchlorate, thulium iii perchlorate, perchloric acid,thulium 3+ salt 8ci,9ci, acmc-1bs9x, perchloric acid thulium iii salt, thulium 3+ triperchlorate, thulium iii perchlorate w/w aqueous solution, reagent grade
PubChem CID 15335706
IUPAC Name thulium(3+);triperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Tm+3]
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11480281
View Documents
Alfa Aesar™
040565.NA
2 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

14014-10-9
467.272
JNNWMYZIHRZEKX-UHFFFAOYSA-K
15335706
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Tm+3]
Cl3O12Tm
MFCD00049609
thulium 3+ perchlorate, thulium iii perchlorate, perchloric acid,thulium 3+ salt 8ci,9ci, acmc-1bs9x, perchloric acid thulium iii salt, thulium 3+ triperchlorate, thulium iii perchlorate w/w aqueous solution, reagent grade
thulium(3+);triperchlorate

Specifications

14014-10-9
Cl3O12Tm
UN3211
JNNWMYZIHRZEKX-UHFFFAOYSA-K
thulium(3+);triperchlorate
15335706
Reagent
Odorless
Thulium(III) perchlorate
Liquid
MFCD00049609
thulium 3+ perchlorate, thulium iii perchlorate, perchloric acid,thulium 3+ salt 8ci,9ci, acmc-1bs9x, perchloric acid thulium iii salt, thulium 3+ triperchlorate, thulium iii perchlorate w/w aqueous solution, reagent grade
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Tm+3]
467.272
467.28
50% w/w aq. soln.
1.51g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.