missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thulium(III) sulfate octahydrate, 99.99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used as a reactant in dehydration reactions. It is the important dopant for fiber amplifiers.
Solubility
Soluble in water, moderately soluble in strong mineral acids.
Notes
Store at room temperature. Keep away from strong oxidizing agents and moisture.
It is used as a reactant in dehydration reactions. It is the important dopant for fiber amplifiers.
Solubility
Soluble in water, moderately soluble in strong mineral acids.
Notes
Store at room temperature. Keep away from strong oxidizing agents and moisture.
Specifications
Specifications
Quantity | 1 g |
Solubility Information | Soluble in water,moderately soluble in strong mineral acids. |
Formula Weight | 770.17 (626.06 Anhydrous) |
Percent Purity | 99.99% |
Sensitivity | Hygroscopic |
Chemical Name or Material | Thulium(III) sulfate octahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'