Learn More
Sampling Systems™ Thumb Tube Pipettes
BIOPHARMA
Economical solution to sampling low viscosity liquids. Sampling Systems™ Thumb Tube Pipettes are ideal for use with a wide range of chemicals.
Brand:  Sampling Systems™ 5055A-50
Product Code. 15859341
  • 121.00 € / Pack of 50
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured with HDPE
  • Individually bagged
  • FDA conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Available sterilized by gamma
  • Press thumb onto the tube until you are at the sampling depth
  • Raise the thumb until levels equalise
  • Press thumb onto the tube to seal the sample in
Specifications
Specifications
HDPE
60 mL
1000 x 12 mm
Individually Bagged
Single-use