Learn More
Thermo Scientific Chemicals Thymidine-5'-monophosphate disodium salt
Description
Thymidine-5'-monophosphate disodium salt is a nucleoside monophosphate used to prepare thymidine diphosphate by thymidylate kinase (TMPK) which is a nucleoside monophosphate kinase. In addition to this, it serves as a constituent of deoxyribonucleic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Thymidine-5′-monophosphate disodium salt is a nucleoside monophosphate used to prepare thymidine diphosphate by thymidylate kinase (TMPK) which is a nucleoside monophosphate kinase. In addition to this, it serves as a constituent of deoxyribonucleic acid.
Solubility
Soluble in water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Specifications
Specifications
Color | White |
Quantity | 100 mg |
Beilstein | 4086927 |
Solubility Information | Soluble in water. |
Formula Weight | 366.18 |
Physical Form | Powder |
Chemical Name or Material | Thymidine-5'-monophosphate disodium salt |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
W27K011 | Certificate of Analysis | 07/10/2024 | J60747.03,J60747.MD,J60747.MC |
S27K003 | Certificate of Analysis | 09/07/2024 | J60747.03,J60747.MD,J60747.MC |
Q11K024 | Certificate of Analysis | 11/06/2024 | J60747.03,J60747.MD,J60747.MC |
Y14I045 | Certificate of Analysis | 29/11/2022 | J60747.03,J60747.MD,J60747.06,J60747.MC |
Y07I043 | Certificate of Analysis | 16/11/2022 | J60747.03,J60747.MD,J60747.06,J60747.MC |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.