missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Thymidine, 99% Product Code.: 11400403

Thermo Scientific Chemicals Thymidine, 99%

Product Code. 11400403
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11400403

Brand: Thermo Scientific Chemicals A11493.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thymidine is used in the syntheses of active pharmaceutical ingredient such as zidovudine. It also pairs with deoxyadenosine in double-stranded deoxyribonucleic acid. It is used to synchronize the cells in G1/early S phase in cell biology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thymidine is used in the syntheses of active pharmaceutical ingredient such as zidovudine. It also pairs with deoxyadenosine in double-stranded deoxyribonucleic acid. It is used to synchronize the cells in G1/early S phase in cell biology.

Solubility
Soluble in water, methanol, hot ethanol, hot acetone, hot ethyl acetate, pyridine, glacial acetic acid and hydrochloric acid. Slightly soluble in hot chloroform .

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-89-5
Molecular Formula C10H14N2O5
Molecular Weight (g/mol) 242.23
MDL Number MFCD00006537
InChI Key IQFYYKKMVGJFEH-UHFFFAOYNA-N
Synonym thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside
PubChem CID 5789
ChEBI CHEBI:17748
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O

Specifications

Melting Point 186°C to 190°C
Odor Odorless
Quantity 5 g
Beilstein 89285
Merck Index 14,9397
Solubility Information Soluble in water,methanol,hot ethanol,hot acetone,hot ethyl acetate,pyridine,glacial acetic acid and hydrochloric acid. Slightly soluble in hot chloroform .
Optical Rotation +19° (c=1 in water)
Formula Weight 242.23
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Thymidine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250677 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA11493.22,A11493.06,A11493.14
Lot Number10247923 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog NumberA11493.22,A11493.06,A11493.14
Lot Number10245366 Certificate TypeCertificate of Analysis Date16/01/2023 Catalog NumberA11493.22,A11493.06,A11493.14
Lot Number10239883 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA11493.22,A11493.06,A11493.14
Lot NumberI8763A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11493.22,A11493.06,A11493.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Thymidine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.