missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Thymidine, 99+% Product Code.: 11986811

Thermo Scientific Chemicals Thymidine, 99+%

Product Code. 11986811
100 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11986811

Brand: Thermo Scientific Chemicals 226741000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-89-5
Molecular Formula C10H14N2O5
Molecular Weight (g/mol) 242.23
InChI Key IQFYYKKMVGJFEH-UHFFFAOYNA-N
Synonym thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside
PubChem CID 5789
ChEBI CHEBI:17748
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O

Specifications

Absorbance 0.62 to 0.68 (A250/A260),0.70 to 0.76 (A280/A260)
Melting Point 187.0°C to 189.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Merck Index 159552
Specific Rotation Condition + 18.50 (20.00°C c=3,H2O)
Specific Rotation + 18.50
Solubility Information Solubility in water: 46g/L (20°C). Other solubilities: soluble in glacial acetic acid, soluble in methanol, hot ethanol, hot acetone, hot, ethyl acetate and pyridine, sparingly soluble in hot chloroform
Spectrophotometry 98.0 to 102% (lambda max. 267nm,pH 7) molar absorptivity 9600+/-4%,on dry basis
Formula Weight 242.23
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Thymidine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0442997 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number226741000,226740250,226740050
Lot NumberA0424738 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number226741000,226740250,226740050
Lot NumberA0396571 Certificate TypeCertificate of Analysis Date16/12/2022 Catalog Number226741000,226740250,226740050
Lot NumberA0399571 Certificate TypeCertificate of Analysis Date16/12/2022 Catalog Number226741000,226740250,226740050
Lot NumberA0394870 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number226741000,226740250,226740050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Thymidine, 99+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.