missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Thymol Blue, Indicator Product Code.: 10474015

Thermo Scientific Chemicals Thymol Blue, Indicator

Product Code. 10474015
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10474015

Brand: Thermo Scientific Chemicals 151450050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-61-9
Molecular Formula C27H30O5S
Molecular Weight (g/mol) 466.592
MDL Number MFCD00005869
InChI Key PRZSXZWFJHEZBJ-UHFFFAOYSA-N
Synonym TB, Thymolsulfonephthalein
PubChem CID 65565
IUPAC Name 4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O

Specifications

Melting Point 221.0°C
Merck Index 15, 9555
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diluted alkali solutions, soluble in alcohol
Infrared Spectrum Authentic
Formula Weight 466.58
Grade Pure
Packaging Glass bottle
Specific Extinction 700 min. (596nm, in 0.01N NaOH)
Visual Transition Interval Acid range: from pH 1.2 (red) to pH 2.8 (yellow), alkaline range: from pH 8.0 (yellow) to pH 9.6 (blue)
Color Brown to Green
Quantity 5 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Thymol Blue
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0457334 Certificate TypeCertificate of Analysis Date10/11/2023 Catalog Number151451000,151450250,151450050
Lot NumberA0402508 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number151451000,151450250,151450050
Lot NumberA0395601 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number151451000,151450250,151450050
Lot NumberA0414196 Certificate TypeCertificate of Analysis Date14/06/2022 Catalog Number151451000,151450250,151450050
Lot NumberA0413175 Certificate TypeCertificate of Analysis Date28/04/2022 Catalog Number151451000,151450250,151450050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Thymol Blue, Indicator >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.