missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Thymol Blue Sodium Salt, For analysis ACS
48.44 € - 285.42 €
Chemical Identifiers
CAS | 62625-21-2 |
---|---|
Molecular Formula | C27H29NaO5S |
Molecular Weight (g/mol) | 488.57 |
MDL Number | MFCD00151093 |
InChI Key | OCMIKNBSIRSUPI-FRWNMSGJSA-M |
PubChem CID | 23692293 |
SMILES | [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10519292
|
Thermo Scientific Chemicals
420940100 |
10 g | Glass bottle |
48.44 €
10g |
In Stock
Log in to see stock. |
||||
10647581
|
Thermo Scientific Chemicals
420940250 |
25 g | Glass bottle |
92.46 €
25g |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||||
10502184
|
Thermo Scientific Chemicals
420941000 |
100 g | Glass bottle |
285.42 €
100g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
62625-21-2 | |
488.57 | |
OCMIKNBSIRSUPI-FRWNMSGJSA-M | |
[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O |
C27H29NaO5S | |
MFCD00151093 | |
23692293 |
Specifications
283.0°C | |
Passes Test | |
MFCD00151093 | |
Authentic | |
sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate | |
23692293 | |
ACS Reagent | |
Black/Brown or Black/Green to Orange | |
Thymol Blue sodium salt |
62625-21-2 | |
C27H29NaO5S | |
OCMIKNBSIRSUPI-FRWNMSGJSA-M | |
[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O | |
488.57 | |
488.58 | |
Acid range: from pH 1.2 (pink) to pH 2.8 (yellow) Alkaline range: from pH 8.0 (yellow) to pH 9.2 (blue) | |
Powder |
Safety and Handling
Product Identifier
- Thymol Blue sodium salt
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title