missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Thymolphthalein
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Thymolphthalein acts as pH indicator, as reagent for blood after decolorizing the alkaline solution by boiling with zinc dust.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thymolphthalein acts as pH indicator, as reagent for blood after decolorizing the alkaline solution by boiling with zinc dust.
Solubility
Soluble in alcohol, acetone and also soluble in dilute alkalis. insoluble in water.
Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
Thymolphthalein acts as pH indicator, as reagent for blood after decolorizing the alkaline solution by boiling with zinc dust.
Solubility
Soluble in alcohol, acetone and also soluble in dilute alkalis. insoluble in water.
Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | 251°C to 253°C |
Quantity | 50 g |
Beilstein | 359413 |
Merck Index | 14,9401 |
Solubility Information | Soluble in alcohol,acetone and also soluble in dilute alkalis. insoluble in water. |
Formula Weight | 430.55 |
Chemical Name or Material | Thymolphthalein |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'