missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Thymolphthalein, Indicator, Pure
52.06 € - 514.56 €
Chemical Identifiers
CAS | 125-20-2 |
---|---|
Molecular Formula | C28H30O4 |
Molecular Weight (g/mol) | 430.544 |
MDL Number | MFCD00005909 |
InChI Key | LDKDGDIWEUUXSH-UHFFFAOYSA-N |
Synonym | TP |
PubChem CID | 31316 |
IUPAC Name | 3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one |
SMILES | CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10418310
|
Thermo Scientific Chemicals
151460100 |
10 g | Glass bottle |
52.06 €
10g |
In Stock
Log in to see stock. |
||||
10795811
|
Thermo Scientific Chemicals
151460500 |
50 g | Glass bottle |
178.89 €
50g |
In Stock
Log in to see stock. |
||||
10328603
|
Thermo Scientific Chemicals
151462500 |
250 g | Glass bottle |
514.56 €
250g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
125-20-2 | |
430.544 | |
LDKDGDIWEUUXSH-UHFFFAOYSA-N | |
31316 | |
CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O |
C28H30O4 | |
MFCD00005909 | |
TP | |
3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one |
Specifications
251.0°C to 253.0°C | |
C28H30O4 | |
18, I, 381 | |
TP | |
LDKDGDIWEUUXSH-UHFFFAOYSA-N | |
CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O | |
430.544 | |
430.53 | |
Pure | |
White | |
Thymolphthalein, Indicator |
125-20-2 | |
MFCD00005909 | |
15, 9556 | |
Solubility in water: insoluble. Other solubilities: soluble in dilute alkali and acids, soluble in alcohol and acetone | |
Authentic | |
3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one | |
31316 | |
1% max. | |
from pH 8.8 (colorless) to pH 10.5 (blue) | |
Fine Crystalline Powder |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title