missing translation for 'onlineSavingsMsg'
Learn More

Manutan Tight head steel drum, lacquered, 210 litre capacity

Brand:  Manutan A350864

Encompass_Preferred

Product Code. 12853809

  • 161.17 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Suitable for the storage of liquids. All drums UN approved and have 1 x 51mm and 1 x 19mm tri-sure fitting.
  • Capacity : 210L
  • Dimensions (H x D) : 880mm x 595mm
  • UN specification : 1A1/X
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers