missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM22 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14927-1-AP-20UL
This item is not returnable.
View return policy
Description
TIM22 is a translocase localized in mitochondrial inner membrane. It is implicated in import of nuclear-encoded mitochondrial preproteins to the mitochondria. TIM22 is the essential core of protein insertion complex (TIM22 complex) that consists of TIM22, TIM54, TIM18, and TIM12.Specifications
TIM22 | |
Polyclonal | |
Unconjugated | |
TIMM22 | |
Testis expressed sequence 4, TEX4, TIM22, TIMM22 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29928, 56322, 79463 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9CQ85, Q9JKW1, Q9Y584 | |
TIMM22 | |
TIM22 Fusion Protein Ag6689 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |