missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Timolol maleate
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Quantity | 1 g |
Solubility | Solubility aq. soln.: soluble. Other solubilities: practically insoluble in ether, soluble in dmso, methanol, ethanol |
Melting Point | 202.0°C to 203.0°C |
Formula Weight | 432.49 |
InChI Key | WLRMANUAADYWEA-NWASOUNVSA-N |
SMILES | OC(=O)\C=C/C(O)=O.CC(C)(C)NC[C@H](O)COC1=NSN=C1N1CCOCC1 |
IUPAC Name | (2S)-1-(tert-butylamino)-3-{[4-(morpholin-4-yl)-1,2,5-thiadiazol-3-yl]oxy}propan-2-ol; (2Z)-but-2-enedioic acid |
Molecular Weight (g/mol) | 432.49 |
Chemical Name or Material | Timolol maleate |
Safety and Handling
Product Identifier
- Timolol maleate
- Warning
- Acute toxicity Category 4
- Reproductive toxicity Category 2
- H302-Harmful if swallowed.
- H361-Suspected of damaging fertility or the unborn child.
- P201-Obtain special instructions before use.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : (S)-3-[3-(tert-Butylamino)-2-hydroxypropoxy]-4-morpholino-1,2,5-thiadiazole monomaleate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'type03FormHeader'
missing translation for 'productTitle'